Skip to Content
MilliporeSigma
All Photos(1)

Documents

726559

Sigma-Aldrich

(S)-(+)-3,3-Dimethyl-2-butylamine

ChiPros®, produced by BASF, 99%

Synonym(s):

(S)-3,3-Dimethyl-2-aminobutane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
((CH3)3C)CH(NH2)CH3
CAS Number:
Molecular Weight:
101.19
Beilstein:
4652328
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

grade

produced by BASF

Assay

≥98.5% (GC)
99%

form

liquid

optical purity

enantiomeric excess: ≥98.5%

refractive index

n20/D 1.413

bp

103 °C

mp

-4 °C

density

0.743 g/mL at 25 °C

SMILES string

C[C@H](N)C(C)(C)C

InChI

1S/C6H15N/c1-5(7)6(2,3)4/h5H,7H2,1-4H3/t5-/m0/s1

InChI key

DXSUORGKJZADET-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ChiPros is a registered trademark of BASF SE

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service