Skip to Content
MilliporeSigma
(S)-(+)-3,3-Dimethyl-2-butylamine 97%

(S)-(+)-3,3-Dimethyl-2-butylamine

(S)-(+)-3,3-Dimethyl-2-butylamine 97%
Synonym(s):
(S)-3,3-Dimethyl-2-aminobutane
Linear Formula:
((CH3)3C)CH(NH2)CH3
CAS Number:
Molecular Weight:
101.19

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing