Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

725854

Sigma-Aldrich

Ethyl 5-(trifluoromethyl)-pyridine-2-carboxylate

≥95.0%

Synonym(s):

Ethyl 5-(trifluoromethyl)picolinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8F3NO2
CAS Number:
Molecular Weight:
219.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥95.0% (GC)
≥95.0%

form

lumps

SMILES string

CCOC(=O)c1ccc(cn1)C(F)(F)F

InChI

1S/C9H8F3NO2/c1-2-15-8(14)7-4-3-6(5-13-7)9(10,11)12/h3-5H,2H2,1H3

InChI key

KCMOTZFSBYHKCI-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service