Skip to Content
MilliporeSigma
All Photos(1)

Documents

718106

Sigma-Aldrich

Isopropyl-1,1,1,3,3,3-d6-amine

99 atom % D

Synonym(s):

2-Aminopropane-d6

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3CH(NH2)CD3
Molecular Weight:
65.15
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

isotopic purity

99 atom % D

Assay

99% (CP)

form

liquid

bp

33-34 °C

density

0.762 g/mL at 25 °C

mass shift

M+6

SMILES string

[2H]C([2H])([2H])C(N)C([2H])([2H])[2H]

InChI

1S/C3H9N/c1-3(2)4/h3H,4H2,1-2H3/i1D3,2D3

InChI key

JJWLVOIRVHMVIS-WFGJKAKNSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-0.4 °F - closed cup

Flash Point(C)

-18 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service