Skip to Content
MilliporeSigma
All Photos(1)

Documents

485497

Sigma-Aldrich

Aniline-13C6

99 atom % 13C

Synonym(s):

13C Labeled aniline, Aminobenzene-13C6, Benzeneamine-13C6, Phenylamine-13C6

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13C6H5NH2
CAS Number:
Molecular Weight:
99.08
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C

Quality Level

Assay

99% (CP)

technique(s)

protein expression: suitable

refractive index

n20/D 1.586 (lit.)

bp

184 °C (lit.)

mp

−6 °C (lit.)

density

1.087 g/mL at 25 °C

mass shift

M+6

storage temp.

2-8°C

SMILES string

N[13c]1[13cH][13cH][13cH][13cH][13cH]1

InChI

1S/C6H7N/c7-6-4-2-1-3-5-6/h1-5H,7H2/i1+1,2+1,3+1,4+1,5+1,6+1

InChI key

PAYRUJLWNCNPSJ-IDEBNGHGSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Muta. 2 - Skin Sens. 1 - STOT RE 1

Target Organs

Blood

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephanie C Morriss et al.
Annals of botany, 120(6), 911-922 (2017-09-30)
Enzymes belonging to the RNase T2 family are essential for normal rRNA turnover in eukaryotes. In Arabidopsis thaliana, this function is performed by RNS2. The null mutant rns2-2 has increased rRNA half-life and constitutive autophagy. The aim of this work

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service