Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

716782

Sigma-Aldrich

N-((Diisopropylamino)methylene)-N-diisopropylaminium tetrafluoroborate

97%

Synonym(s):

N,N,N′,N′-Tetraisopropylformamidinium tetrafluoroborate, Bis-(Diisopropylamino)methylium tetrafluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H29BF4N2
CAS Number:
Molecular Weight:
300.19
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

reagent type: catalyst

mp

280-284 °C

SMILES string

F[B-](F)(F)F.CC(C)N(\C=[N+](\C(C)C)C(C)C)C(C)C

InChI

1S/C13H29N2.BF4/c1-10(2)14(11(3)4)9-15(12(5)6)13(7)8;2-1(3,4)5/h9-13H,1-8H3;/q+1;-1

InChI key

PEKVUVXZBLFOHV-UHFFFAOYSA-N

Application

Reactant for cross-coupling reactions

Legal Information

Sold under license from Kanata Chemical Technologies Inc. for research purposes only.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bhartesh Dhudshia et al.
Chemical communications (Cambridge, England), 6(6), 668-670 (2006-02-01)
Acyclic diaminocarbenes are found to be useful ligands for palladium catalyzed Suzuki-Miyaura, Sonogashira and Heck cross-coupling reactions of aryl/alkenyl bromides and chlorides.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service