Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

659983

Sigma-Aldrich

1,3-Di-tert-butylimidazolium tetrafluoroborate

97%

Synonym(s):

1,3-Bis(tert-butyl)-imidazol-2-ylidinium tetrafluoroborate, N,N′-Bis(tert-butyl)imidazolium tetrafluoroborate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$95.90
5 G
$327.00

$95.90


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$95.90
5 G
$327.00

About This Item

Empirical Formula (Hill Notation):
C11H21N2 · BF4
CAS Number:
Molecular Weight:
268.10
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$95.90


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

reaction suitability

reagent type: catalyst

mp

157-198 °C

SMILES string

F[B-](F)(F)F.CC(C)(C)n1cc[n+](c1)C(C)(C)C

InChI

1S/C11H21N2.BF4/c1-10(2,3)12-7-8-13(9-12)11(4,5)6;2-1(3,4)5/h7-9H,1-6H3;/q+1;-1

InChI key

OOFLHRYFPBGTPQ-UHFFFAOYSA-N

General description

1,3-Di-tert-butylimidazolium tetrafluoroborate is an N-heterocyclic carbene (NHC) compound that can be prepared by reacting paraformaldehyde with tert-butyl amine and hydrogen tetrafluoroborate (35% in water).[1]

Application

1,3-Di-tert-butylimidazolium tetrafluoroborate may be used in the preparation of di-μ-iodobis(1,3-di-tert-butylimidazolin-2-ylidene)diiododipalladium(II), a palladium(II)-NHC complex, which can catalyze Mizoroki-Heck reaction.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, structure and catalytic application of palladium (II) complexes bearing N-heterocyclic carbenes and phosphines.
Herrmann WA, et al.
Journal of Organometallic Chemistry, 617, 616-628 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service