Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

709026

Sigma-Aldrich

Dirhodium tetracaprolactamate

97%

Synonym(s):

Rh2(cap)4, Tetrakis[μ-(hexahydro-2H-azepin-2-onato-κN1,κO2)]dirhodium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H40N4O4Rh2
CAS Number:
Molecular Weight:
654.41
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

core: rhodium
reagent type: catalyst

mp

269-274 °C (lit.)

SMILES string

[Rh][Rh].O=C1CCCCCN1.O=C2CCCCCN2.O=C3CCCCCN3.O=C4CCCCCN4

InChI

1S/4C6H11NO.2Rh/c4*8-6-4-2-1-3-5-7-6;;/h4*1-5H2,(H,7,8);;

InChI key

SSCZKSBMOZFVCQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for selective carbenoid reactions of diazo compounds

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service