Skip to Content
MilliporeSigma
All Photos(1)

Documents

725455

Sigma-Aldrich

Rhodium(II) triphenylacetate dimer

as complex with dichloromethane

Synonym(s):

Rh2(TPA)4, Rh2(triphenylacetate)4, Rhodiumtriphenylacetate, Tetrakis(triphenylacetato)dirhodium(II)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C80H60O8Rh2 · CH2Cl2
CAS Number:
Molecular Weight:
1440.07
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

core: rhodium
reagent type: catalyst
reaction type: C-H Activation

mp

260-263 °C

SMILES string

ClCCl.O=C(O[Rh](OC(=O)C(c1ccccc1)(c2ccccc2)c3ccccc3)[Rh](OC(=O)C(c4ccccc4)(c5ccccc5)c6ccccc6)OC(=O)C(c7ccccc7)(c8ccccc8)c9ccccc9)C(c%10ccccc%10)(c%11ccccc%11)c%12ccccc%12

InChI

1S/4C20H16O2.CH2Cl2.2Rh/c4*21-19(22)20(16-10-4-1-5-11-16,17-12-6-2-7-13-17)18-14-8-3-9-15-18;2-1-3;;/h4*1-15H,(H,21,22);1H2;;/q;;;;;2*+2/p-4

InChI key

SRAMLXDHXNCYQM-UHFFFAOYSA-J

Application

Catalyst for:
  • Amination of allene carbamates
  • Allene aziridination to stereoselectively prepare cyclic cabamates
  • C-H bond nitrene insertion
  • Enantioselective cyclopropanation
  • Preparation of cyclic oxonium ylides

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service