Skip to Content
MilliporeSigma
All Photos(1)

Documents

701432

Sigma-Aldrich

L-Threonine-2,3,4,4,4-d5

98 atom % D, 97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3CD(OH)CD(NH2)CO2H
CAS Number:
Molecular Weight:
124.15
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

97% (CP)

form

powder

mp

256 °C (dec.)

mass shift

M+5

SMILES string

[2H]C([2H])([2H])[C@@]([2H])(O)[C@]([2H])(N)C(O)=O

InChI

1S/C4H9NO3/c1-2(6)3(5)4(7)8/h2-3,6H,5H2,1H3,(H,7,8)/t2-,3+/m1/s1/i1D3,2D,3D

InChI key

AYFVYJQAPQTCCC-WKHZCAFQSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service