Skip to Content
MilliporeSigma
All Photos(1)

Documents

392901

Sigma-Aldrich

N,N-Dimethylformamide-d

98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
DCON(CH3)2
CAS Number:
Molecular Weight:
74.10
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

form

liquid

refractive index

n20/D 1.427 (lit.)

bp

153 °C (lit.)

mp

-61 °C (lit.)

density

0.957 g/mL at 25 °C

mass shift

M+1

SMILES string

[2H]C(=O)N(C)C

InChI

1S/C3H7NO/c1-4(2)3-5/h3H,1-2H3/i3D

InChI key

ZMXDDKWLCZADIW-WFVSFCRTSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of Labelled Compounds & Radiopharmaceuticals, 34, 263-263 (1994)
Claudia Orelio et al.
Haematologica, 94(3), 409-413 (2009-02-13)
Shwachman-Diamond syndrome is a hereditary disorder characterized by pancreatic insufficiency and bone marrow failure. Most Shwachman-Diamond syndrome patients have mutations in the SBDS gene located at chromosome 7 and suffer from recurrent infections, due to neutropenia in combination with impaired
Bart R B Haverkorn van Rijsewijk et al.
Molecular systems biology, 7, 477-477 (2011-04-01)
Despite our increasing topological knowledge on regulation networks in model bacteria, it is largely unknown which of the many co-occurring regulatory events actually control metabolic function and the distribution of intracellular fluxes. Here, we unravel condition-dependent transcriptional control of Escherichia

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service