Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

701017

Sigma-Aldrich

2-Furanylboronic acid MIDA ester

97%

Synonym(s):

2-(Furan-2-yl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione, 2-Furanboronic acid MIDA ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10BNO5
CAS Number:
Molecular Weight:
222.99
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

97%

form

powder

mp

130-135 °C

SMILES string

CN1CC(=O)OB(OC(=O)C1)c2ccco2

InChI

1S/C9H10BNO5/c1-11-5-8(12)15-10(16-9(13)6-11)7-3-2-4-14-7/h2-4H,5-6H2,1H3

InChI key

QNZYBMSGFWTQOS-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David M Knapp et al.
Journal of the American Chemical Society, 131(20), 6961-6963 (2009-05-02)
Many boronic acids, including 2-heterocyclic, vinyl, and cyclopropyl derivatives, are inherently unstable, which can limit their benchtop storage and/or efficient cross-coupling. We herein report the first general solution to this problem: in situ slow release of unstable boronic acids from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service