Skip to Content
MilliporeSigma
All Photos(1)

Documents

699004

Sigma-Aldrich

L-Ascorbic acid-2-13C

≥99 atom % 13C, ≥98% (CP)

Synonym(s):

L-Threoascorbic acid-2-13C, Antiscorbutic factor-2-13C, Vitamin C-2-13C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC5H8O6
CAS Number:
Molecular Weight:
177.12
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

≥99 atom % 13C

Assay

≥98% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

color

white to off-white

mp

190-194 °C (dec) (lit.)

mass shift

M+1

SMILES string

OC[C@H](O)[C@H]1OC(=O)[13C](O)=C1O

InChI

1S/C6H8O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2,5,7-10H,1H2/t2-,5+/m0/s1/i4+1

InChI key

CIWBSHSKHKDKBQ-DXEGWCNNSA-N

General description

L-Ascorbic acid or vitamin C is a water-soluble vitamin found in various food items. It is useful in the biosynthesis of collagen, L-carnitine, and neurotransmitters. Vitamin C is also function as an antioxidant.

Application

L-Ascorbic acid-2-13C (vitamin C-2-13C) is also known as L-threoascorbic acid-2-13C. This stable isotope internal standard is suitable for the quantitation of L-ascorbic acid in dietary supplements, fortified foods or biological fluids using a variety of testing applications.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vitamin c
Linster CL and Van SE
FEBS Journal, 274(1), 1-22 (2007)
A review of micronutrients in sepsis: the role of thiamine, l-carnitine, vitamin C, selenium and vitamin D
Belsky JB, et al.
Nutrition Research Reviews, 31(2), 281-290 (2018)
First bioarchaeological evidence of probable scurvy in Southeast Asia: multifactorial etiologies of vitamin C deficiency in a tropical environment
Halcrow SE, et al.
International Journal of Paleopathology, 5(1), 63-71 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service