Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V-047

Supelco

L-Ascorbic Acid (Vitamin C)-13C6 solution

500 μg/mL in acetonitrile: water, certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C6H8O6
Molecular Weight:
182.08
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

500 μg/mL in acetonitrile: water

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−70°C

SMILES string

O1[C@@H](C(C(=O)C1=O)O)[C@@H](O)CO

InChI

1S/C6H8O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2-3,5,7-9H,1H2/t2-,3?,5+/m0/s1

InChI key

KRGQEOSDQHTZMX-IGCYCDGOSA-N

General description

Serum levels of L-ascorbic acid, commonly known as vitamin C, are an adequate measurement of clinical status. Low levels of vitamin C occur in scurvy, malabsorption, alcoholism, pregnancy, hyperthyroidism, and renal failure.

Application

L-Ascorbic Acid (Vitamin C)-13C6 solution, (13C6-ascorbic acid) may be used as a certified reference material (CRM) for the determination of ascorbate in aqueous solutions and plasma by gas chromatography-mass spectrometry.
This stable-labeled internal standard is suitable for quantification of vitamin C levels in serum or other matrices by LC/MS or GC/MS for clinical diagnostic, or forensic analysis applications.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ascorbate and dehydroascorbate measurements in aqueous solutions and plasma determined by gas chromatography-mass spectrometry
Deutsch JC and Kolhouse JF
Analytical Chemistry, 65(4), 321-326 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service