Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

693383

Sigma-Aldrich

cBRIDP

Synonym(s):

Di-tert-butyl(2,2-diphenyl-1-methyl-1-cyclopropyl)phosphine, Mo-Phos

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H33P
CAS Number:
Molecular Weight:
352.49
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

form

crystals

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

87-91 °C

functional group

phosphine

SMILES string

CC(C)(C)P(C(C)(C)C)C1(C)CC1(c2ccccc2)c3ccccc3

InChI

1S/C24H33P/c1-21(2,3)25(22(4,5)6)23(7)18-24(23,19-14-10-8-11-15-19)20-16-12-9-13-17-20/h8-17H,18H2,1-7H3

InChI key

QMLPJDVGNRHGJQ-UHFFFAOYSA-N

Legal Information

Sold in collaboration with Takasago for research purposes only.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Hydrogenation, Asymmetric Catalysis, Binap, SEGPHOS®, Aldol reaction, Alkenylation, Arylation, Mannich reaction, Fluorination, Michael addition, Hydrosilylation, Cycloaddition, Takasago

Protocols

TPGS-750-M surfactant enables various reactions in water at room temperature, enhancing efficiency and versatility in synthesis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service