Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

711179

Sigma-Aldrich

vBRIDP

97%

Synonym(s):

Bis(1,1-dimethylethyl)(1-methyl-2,2-diphenylethenyl)phosphine, Di-tert-butyl(1-methyl-2,2-diphenylethenyl)phosphine

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$108.00

$108.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$108.00

About This Item

Empirical Formula (Hill Notation):
C23H31P
CAS Number:
Molecular Weight:
338.47
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

$108.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

97%

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

130-133 °C

functional group

phosphine

storage temp.

2-8°C

SMILES string

C\C(P(C(C)(C)C)C(C)(C)C)=C(/c1ccccc1)c2ccccc2

InChI

1S/C23H31P/c1-18(24(22(2,3)4)23(5,6)7)21(19-14-10-8-11-15-19)20-16-12-9-13-17-20/h8-17H,1-7H3

InChI key

CYGZMOQFIMJEIT-UHFFFAOYSA-N

Application

Catalyst for:
  • Preparation of biphenyls
  • Amination of aryl bromides and chlorides

Legal Information

Sold in collaboration with Takasago for research purposes only. US6455720

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Hydrogenation, Asymmetric Catalysis, Binap, SEGPHOS®, Aldol reaction, Alkenylation, Arylation, Mannich reaction, Fluorination, Michael addition, Hydrosilylation, Cycloaddition, Takasago

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service