Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

691496

Sigma-Aldrich

Borane di(tert-butyl)phosphine complex

97%

Synonym(s):

(Di-tert-butylphosphine)trihydroboron, Di-tert-butylphosphine compound with borane (1:1), Di-tert-butylphosphine borane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H19P · BH3
CAS Number:
Molecular Weight:
160.05
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22
To inquire about this product 691496, please contact your local MilliporeSigma office or dealer. Contact Technical Service

Assay

97%

form

solid

reaction suitability

reagent type: reductant

mp

59-64 °C

functional group

phosphine

SMILES string

B.CC(C)(C)PC(C)(C)C

InChI

1S/C8H19P.BH3/c1-7(2,3)9-8(4,5)6;/h9H,1-6H3;1H3

InChI key

JPINHYFOWCBINQ-UHFFFAOYSA-N

Application

Reactant for:
  • Preparation of N-heterocyclic carbene borane complexes by Lewis base exchange reactions
  • Dehydrocoupling reactions
  • Tele-substitution reactions on fluorobenzenechromium complexes
  • Palladium-catalyzed biaryl-coupling reactions
  • Palladium-catalyzed Suzuki coupling

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service