Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

677647

Sigma-Aldrich

tert-Butyldimethylphosphine borane

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$275.00

$275.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$275.00

About This Item

Linear Formula:
C6H15P · BH3
CAS Number:
Molecular Weight:
131.99
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

$275.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

159-168 °C

functional group

phosphine

SMILES string

B.CP(C)C(C)(C)C

InChI

1S/C6H15P.BH3/c1-6(2,3)7(4)5;/h1-5H3;1H3

InChI key

MHAGKNYBOOCWIH-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Water-react 2

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service