Skip to Content
MilliporeSigma
All Photos(2)

Documents

687782

Sigma-Aldrich

Indole-2-carboxaldehyde

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7NO
CAS Number:
Molecular Weight:
145.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

138-142 °C

SMILES string

O=Cc1cc2ccccc2[nH]1

InChI

1S/C9H7NO/c11-6-8-5-7-3-1-2-4-9(7)10-8/h1-6,10H

InChI key

SBNOTUDDIXOFSN-UHFFFAOYSA-N

Application

Useful starter in indole and natural product chemistry

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical & Pharmaceutical Bulletin, 39, 2170-2170 (1991)
Alexandre A Pletnev et al.
The Journal of organic chemistry, 67(26), 9276-9287 (2002-12-21)
2-iodobenzonitrile, its derivatives, and various heterocyclic analogues undergo palladium(0)-catalyzed annulation onto diarylacetylenes or bicyclic alkenes to afford 2,3-diarylindenones and polycyclic aromatic ketones in very good to excellent yields. This reaction represents one of the first examples of the addition of
Cédric Peyrot et al.
Molecules (Basel, Switzerland), 25(9) (2020-05-10)
Faced with the ban of some organic UV filters such as octinoxate or avobenzone, especially in Hawaii, it became essential to offer new alternatives that are both renewable and safe for humans and the environment. In this context, a class
Niina Tani et al.
Bioorganic & medicinal chemistry, 22(23), 6655-6664 (2014-12-03)
Inhibition of CYP2A6-mediated nicotine metabolism can reduce cigarette smoking. We sought potent and selective CYP2A6 inhibitors to be used as leads for drugs useful in smoking reduction therapy, by evaluating CYP2A6 inhibitory effect of novel formyl, alkyl amine or carbonitrile

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service