MilliporeSigma
  • Carbopalladation of nitriles: synthesis of 2,3-diarylindenones and polycyclic aromatic ketones by the Pd-catalyzed annulation of alkynes and bicyclic alkenes by 2-iodoarenenitriles.
Carbopalladation of nitriles: synthesis of 2,3-diarylindenones and polycyclic aromatic ketones by the Pd-catalyzed annulation of alkynes and bicyclic alkenes by 2-iodoarenenitriles.
The Journal of organic chemistry (2002-12-21)
Alexandre A Pletnev, Qingping Tian, Richard C Larock
ABSTRACT
2-iodobenzonitrile, its derivatives, and various heterocyclic analogues undergo palladium(0)-catalyzed annulation onto diarylacetylenes or bicyclic alkenes to afford 2,3-diarylindenones and polycyclic aromatic ketones in very good to excellent yields. This reaction represents one of the first examples of the addition of an organopalladium moiety to the carbon-nitrogen triple bond of a nitrile. The reaction is compatible with a number of functional groups. A reaction mechanism, as well as a model accounting for the electronic effects of substituents on the aromatic ring of the nitrile, is proposed.