Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

683663

Sigma-Aldrich

Fmoc-α-Me-Ala-OH-15N

99 atom % 15N, 98% (CP)

Synonym(s):

2-Aminoisobutyric acid-15N, α-N-Fmoc

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H1915NO4
CAS Number:
Molecular Weight:
326.35
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

99 atom % 15N

Assay

98% (CP)

form

solid

mol wt

326.35 by atom % calculation

technique(s)

bio NMR: suitable

application(s)

peptide synthesis

functional group

Fmoc

mass shift

M+1

SMILES string

CC(C)([15NH]C(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O

InChI

1S/C19H19NO4/c1-19(2,17(21)22)20-18(23)24-11-16-14-9-5-3-7-12(14)13-8-4-6-10-15(13)16/h3-10,16H,11H2,1-2H3,(H,20,23)(H,21,22)/i20+1

InChI key

HOZZVEPRYYCBTO-UUZXFVOJSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service