Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

683418

Sigma-Aldrich

Lithium dibutyl(isopropyl)magnesate

0.7 M in diethyl ether/hexanes

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H25LiMg
CAS Number:
Molecular Weight:
188.56
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

liquid

reaction suitability

reaction type: Grignard Reaction

concentration

0.7 M in diethyl ether/hexanes

density

0.708 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

[Li+].CCCC[Mg-](CCCC)C(C)C

InChI

1S/2C4H9.C3H7.Li.Mg/c2*1-3-4-2;1-3-2;;/h2*1,3-4H2,2H3;3H,1-2H3;;/q;;;+1;-1

InChI key

NRIMXEIECWMTKG-UHFFFAOYSA-N

General description

Haziness is due to slow hydrolysis of product. Does not effect use.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Skin Irrit. 2 - STOT SE 3 - Water-react 1

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service