Skip to Content
MilliporeSigma
All Photos(1)

Documents

703559

SAFC

Lanthanum(III) chloride bis(lithium chloride) complex solution

0.6 M in THF

Synonym(s):

Lanthanum trichloride lithium chloride complex

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LaCl3 · 2LiCl
CAS Number:
Molecular Weight:
330.05
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

reaction suitability

reagent type: catalyst
core: lanthanum

concentration

0.6 M in THF
16 % (w/w)

density

1.05 g/mL at 25 °C

SMILES string

[Li]Cl.[Li]Cl.Cl[La](Cl)Cl

InChI

1S/5ClH.La.2Li/h5*1H;;;/q;;;;;+3;2*+1/p-5

InChI key

YSKOYVQYUJUXCN-UHFFFAOYSA-I

Related Categories

Application

Selective 1,2-Additions with LaCl3•2LiCl

  • Mediates Grignard additions
  • Reagent for metal exchange reactions
  • Promoter for the addition of organometallic compounds to sterically hindered, enolizable or α,β-unsaturated ketones or imines

Legal Information

Product of Albemarle US Inc

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Skin Sens. 1 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

62.6 °F

Flash Point(C)

17 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Grignard reagent's 1,2-addition to ketones is powerful but selective issues often arise from competitive alpha-deprotonation.

Transformative reagents enable selective conversions within molecules containing sensitive functionalities under mild reactions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service