Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

679399

Sigma-Aldrich

(S)-(+)-2-Aminobutanamide hydrochloride

97%

Synonym(s):

(S)-2-Aminobutyramide hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$115.00

$115.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$115.00

About This Item

Linear Formula:
CH3CH2CH(NH2)CONH2 · HCl
CAS Number:
Molecular Weight:
138.60
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

$115.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

97%

form

solid

optical activity

[α]22/D +24°, c = 1 in H2O

mp

259-263 °C

functional group

amide
amine

SMILES string

CC[C@H](N)C(N)=O.[H]Cl

InChI

1S/C4H10N2O.ClH/c1-2-3(5)4(6)7;/h3H,2,5H2,1H3,(H2,6,7);1H/t3-;/m0./s1

InChI key

HDBMIDJFXOYCGK-DFWYDOINSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service