Skip to Content
MilliporeSigma
(S)-(+)-2-Aminobutanamide hydrochloride 97%

(S)-(+)-2-Aminobutanamide hydrochloride

(S)-(+)-2-Aminobutanamide hydrochloride 97%
Synonym(s):
(S)-2-Aminobutyramide hydrochloride
Linear Formula:
CH3CH2CH(NH2)CONH2 · HCl
CAS Number:
Molecular Weight:
138.60

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing