Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

679356

Sigma-Aldrich

[3-(Diethylamino)propyl]trimethoxysilane

deposition grade, ≥97%

Synonym(s):

N,N-Diethyl-3-(trimethoxysilyl)propylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2N(CH2)3Si(OCH3)3
CAS Number:
Molecular Weight:
235.40
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

grade

deposition grade

Assay

≥97%

form

liquid

refractive index

n20/D 1.423 (lit.)

bp

120 °C/20 mmHg (lit.)

density

0.95 g/mL at 25 °C (lit.)

SMILES string

CCN(CC)CCC[Si](OC)(OC)OC

InChI

1S/C10H25NO3Si/c1-6-11(7-2)9-8-10-15(12-3,13-4)14-5/h6-10H2,1-5H3

InChI key

ZLDHYRXZZNDOKU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Deposition Grade Silanes, fully characterized by chemical analysis and nuclear magnetic resonance (NMR) with greater than 98% purity, for Sol-Gel Processes.

Mesoporous materials, such as aerogels, offer advantages for practical hydrogen storage. They have large surface areas, open porosity, small pore sizes, and the ability to coat the surface with one or more compounds.

atomic layer deposition (ALD), microelectronics, Mo:Al2O3 films, nanocomposite coating, photovoltaics, semiconductor devices, W:Al2O3 films, composite films, layer-by-layer

Silica's versatility spans various industries, including biomedical applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service