Skip to Content
MilliporeSigma
[3-(Diethylamino)propyl]trimethoxysilane 96%

[3-(Diethylamino)propyl]trimethoxysilane

[3-(Diethylamino)propyl]trimethoxysilane 96%
Synonym(s):
N,N-Diethyl-3-(trimethoxysilyl)propylamine
Linear Formula:
(C2H5)2N(CH2)3Si(OCH3)3
CAS Number:
Molecular Weight:
235.40
EC Number:

Products

Boiling Point (°C)
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing