Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

667188

Sigma-Aldrich

2-Bromo-1-tetralone

97%

Synonym(s):

2-Bromo-3,4-dihydro-1(2H)-naphthalenone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9OBr
CAS Number:
Molecular Weight:
225.08
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

bp

64-67 °C/0.06 mmHg

mp

39-43 °C

storage temp.

2-8°C

SMILES string

BrC1CCc2ccccc2C1=O

InChI

1S/C10H9BrO/c11-9-6-5-7-3-1-2-4-8(7)10(9)12/h1-4,9H,5-6H2

InChI key

AYNCWMIFKFADCZ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service