Skip to Content
MilliporeSigma
All Photos(1)

Documents

C32607

Sigma-Aldrich

2-Chlorocyclohexanone

98%

Synonym(s):

α-Chlorocyclohexanone, (±)-2-Chloro-1-cyclohexanone, (±)-2-Chlorocyclohexanone, 2-Chlorocyclohexan-1-one, 2-Chlorocyclohexanone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H9(=O)
CAS Number:
Molecular Weight:
132.59
Beilstein:
774100
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

refractive index

n20/D 1.484 (lit.)

bp

82-83 °C/10 mmHg (lit.)

mp

23 °C (lit.)

density

1.161 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

ClC1CCCCC1=O

InChI

1S/C6H9ClO/c7-5-3-1-2-4-6(5)8/h5H,1-4H2

InChI key

CCHNWURRBFGQCD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Gold et al.
Chemico-biological interactions, 45(3), 305-314 (1983-08-01)
The pseudo-first-order hydrolysis rate constants in pH 7.4 buffer-acetone solution are reported for 1-chlorocyclohexene oxide and 2-chlorocyclohexanone at 0, 25 and 37 degrees C. The rate constants, in conjunction with product studies, demonstrate that the hydrolysis of 1-chlorocyclohexane oxide quantitatively
Patrícia F Monteiro et al.
Journal of controlled release : official journal of the Controlled Release Society, 323, 549-564 (2020-05-07)
Triple negative or basal-like breast cancer (TNBC) is characterised by aggressive progression, lack of standard therapies and poorer overall survival rates for patients. The bad prognosis, high rate of relapse and resistance against anticancer drugs have been associated with a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service