Skip to Content
MilliporeSigma
All Photos(1)

Documents

645672

Sigma-Aldrich

Uridine-13C9,15N2 5′-triphosphate disodium salt solution

100 mM (in 5mM Tris HCl / H2O), ≥98 atom %, ≥95% (CP)

Synonym(s):

13C and 15N Labeled uridine 5′-triphosphate disodium salt solution, UTP-13C9,15N2 disodium salt solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C9H1315N2Na2O15P3
Molecular Weight:
539.03
UNSPSC Code:
41106305
NACRES:
NA.12

isotopic purity

≥98 atom %

Quality Level

Assay

≥95% (CP)

concentration

100 mM (in 5mM Tris HCl / H2O)

technique(s)

bio NMR: suitable

mass shift

M+11

storage temp.

−20°C

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service