Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

636738

Sigma-Aldrich

1,4-Bis(2-ethylhexyl)benzene

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4[CH2CH(C2H5)(CH2)3CH3]2
CAS Number:
Molecular Weight:
302.54
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

refractive index

n20/D 1.4853 (lit.)

bp

190 °C/0.4 mmHg (lit.)

density

0.851 g/mL at 25 °C (lit.)

SMILES string

CCCCC(CC)Cc1ccc(CC(CC)CCCC)cc1

InChI

1S/C22H38/c1-5-9-11-19(7-3)17-21-13-15-22(16-14-21)18-20(8-4)12-10-6-2/h13-16,19-20H,5-12,17-18H2,1-4H3

InChI key

KGDLKOAYUABDIH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maurus Biedermann et al.
Journal of chromatography. A, 1521, 140-149 (2017-09-25)
On-line coupled high performance liquid chromatography-gas chromatography-flame ionization detection (HPLC-GC-FID) is the most widely used method for the analysis of mineral oil hydrocarbons in food, food contact materials, tissues and cosmetics. With comprehensive two-dimensional gas chromatography (GCxGC), a tool became
Maurus Biedermann et al.
Journal of chromatography. A, 1624, 461236-461236 (2020-06-17)
On-line coupled high performance liquid chromatography-gas chromatography-flame ionization detection (HPLC-GC-FID) used for determining mineral oil aromatic hydrocarbons (MOAH) in foods, particularly in certain oils and fats, may be disturbed by interfering olefins present as natural food components or resulting from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service