Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

616877

Sigma-Aldrich

1,2,3,4-Tetramethylcyclopentadiene-2-methyl-d3

98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9D3H11
CAS Number:
Molecular Weight:
125.23
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

85% (CP)

mass shift

M+3

SMILES string

[2H]C([2H])([2H])C1=C(C)CC(C)=C1C

InChI

1S/C9H14/c1-6-5-7(2)9(4)8(6)3/h5H2,1-4H3/i3D3

InChI key

VNPQQEYMXYCAEZ-HPRDVNIFSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

107.6 °F - closed cup

Flash Point(C)

42 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service