Skip to Content
MilliporeSigma
All Photos(1)

Documents

613614

Sigma-Aldrich

Ethane-d5-thiol

98 atom % D

Synonym(s):

Ethyl-d5 mercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3CD2SH
CAS Number:
Molecular Weight:
67.16
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

99% (CP)

technique(s)

protein expression: suitable

mass shift

M+5

storage temp.

−20°C

SMILES string

[2H]C([2H])([2H])C([2H])([2H])S

InChI

1S/C2H6S/c1-2-3/h3H,2H2,1H3/i1D3,2D2

InChI key

DNJIEGIFACGWOD-ZBJDZAJPSA-N

Related Categories

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-49.0 °F - closed cup - (in analogy to similar products)

Flash Point(C)

-45 °C - closed cup - (in analogy to similar products)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service