Skip to Content
MilliporeSigma
All Photos(1)

Documents

600164

Sigma-Aldrich

Methane-13C

10 atom % 13C

Synonym(s):

13C Labeled methane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CH4
CAS Number:
Molecular Weight:
17.04
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

vapor density

0.55 (vs air)

isotopic purity

10 atom % 13C

mol wt

16.13 by atom % calculation (10 atom % 13C)

bp

−161 °C (lit.)

mp

−183 °C (lit.)

mass shift

M+1

SMILES string

[13CH4]

InChI

1S/CH4/h1H4/i1+1

InChI key

VNWKTOKETHGBQD-OUBTZVSYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1A - Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

-306.4 °F - closed cup

Flash Point(C)

-188 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service