Skip to Content
MilliporeSigma
All Photos(1)

Documents

603732

Sigma-Aldrich

Iodobenzene-13C6

99 atom % 13C, 99% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13C6H5I
CAS Number:
Molecular Weight:
209.96
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

99% (CP)

form

solid

bp

188 °C (lit.)

mp

-29 °C (lit.)

density

1.876 g/mL at 25 °C

mass shift

M+6

SMILES string

I[13c]1[13cH][13cH][13cH][13cH][13cH]1

InChI

1S/C6H5I/c7-6-4-2-1-3-5-6/h1-5H/i1+1,2+1,3+1,4+1,5+1,6+1

InChI key

SNHMUERNLJLMHN-IDEBNGHGSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service