596930
Dichlorobis(triphenylphosphine)palladium(II), polymer-bound
200-400 mesh, extent of labeling: 1.0-2.0 mmol/g loading, 2 % cross-linked with divinylbenzene
Synonym(s):
Bis(triphenylphosphine)palladium(II) dichloride, polymer-bound
About This Item
Recommended Products
crosslinking
2 % cross-linked with divinylbenzene
reaction suitability
core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reaction type: solution phase peptide synthesis
reagent type: catalyst
extent of labeling
1.0-2.0 mmol/g loading
particle size
200-400 mesh
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service