Skip to Content
MilliporeSigma
All Photos(3)

Documents

593583

Sigma-Aldrich

Copper-zinc alloy

nanopowder, <150 nm particle size (SEM), 56-60% Cu basis, 37-41% Zn basis

Synonym(s):

Cu34Zn11

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cu34Zn11
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

nanopowder

composition

copper, 60 wt. %
Cu, 56-60%
zinc, 40 wt. %
Zn, 37-41%

particle size

<150 nm (SEM)

cation traces

Mn: 1.5-2.5%
Pb: 0.5-2.5%
Sn: 1.5-2.0%

SMILES string

[Cu].[Zn]

InChI

1S/Cu.Zn

InChI key

TVZPLCNGKSPOJA-UHFFFAOYSA-N

Physical form

Macro-agglomeration of ultra-fine particles, with size of about 70 nanometers.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service