Skip to Content
MilliporeSigma
All Photos(1)

Documents

576824

Sigma-Aldrich

Silver-copper alloy

nanopowder, <100 nm particle size

Synonym(s):

CuAg25

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuAg25
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

nanopowder

composition

Ag, ~97.5%
Cu, ~2.5%

particle size

<100 nm

SMILES string

[Cu].[Ag]

InChI

1S/Ag.Cu

InChI key

NEIHULKJZQTQKJ-UHFFFAOYSA-N

General description

Organically coated to promote dispersion in organic solvents.

Legal Information

Product of Engi-Mat Co.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service