Skip to Content
MilliporeSigma
All Photos(1)

Documents

589977

Sigma-Aldrich

Thiosulfate, polymer-bound

macroporous, 16-50 mesh, extent of labeling: 1.5 mmol/g loading

Synonym(s):

Benzyltrimethylammonium thiosulfate, polymer-bound

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

Quality Level

form

solid

reaction suitability

reaction type: solution phase peptide synthesis

extent of labeling

1.5 mmol/g loading

particle size

16-50 mesh

Related Categories

Features and Benefits

This resin has been reported to successfully quench Dess-Martin and Grieco-Dess-Martin periodinane reagents after oxidation step. Also can be used as a scavenger for allyl and benzyl halides.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Parlow, J.J. et al.
Tetrahedron, 55, 6785-6785 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service