Skip to Content
MilliporeSigma
All Photos(1)

Documents

549363

Sigma-Aldrich

Carbonylimidazole, polymer-bound

100-200 mesh, extent of labeling: 0.7-1.5 mmol/g loading, 1 % cross-linked

Synonym(s):

4-Benzyloxybenzyl imidazole-1-carboxylate polymer-bound, Activated carbonate polymer-bound, Imidazole carbonate, polymer-bound, Imidazole carbonate, polymer-bound to Wang resin, Imidazole carboxylate-Wang resin, Wang imidazolide carbamate resin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Quality Level

crosslinking

1 % cross-linked

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis
reactivity: amine reactive

extent of labeling

0.7-1.5 mmol/g loading

particle size

100-200 mesh

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service