Skip to Content
MilliporeSigma
All Photos(1)

Documents

588199

Sigma-Aldrich

Glyphosate-3-13C

99 atom % 13C

Synonym(s):

N-(phosphonomethyl-13C)-Glycine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH2NH13CH2PO3H2
CAS Number:
Molecular Weight:
170.07
UNSPSC Code:
12352116
PubChem Substance ID:

isotopic purity

99 atom % 13C

mp

230 °C (dec.) (lit.)

application(s)

agriculture
environmental

format

neat

mass shift

M+1

SMILES string

OC(=O)CN[13CH2]P(O)(O)=O

InChI

1S/C3H8NO5P/c5-3(6)1-4-2-10(7,8)9/h4H,1-2H2,(H,5,6)(H2,7,8,9)/i2+1

InChI key

XDDAORKBJWWYJS-VQEHIDDOSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service