Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

586455

2,4-Dinitrophenol-3,5,6-d3

98 atom % D, moist solid; "wetted"

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6D3HN2O5
CAS Number:
Molecular Weight:
187.12
MDL number:
UNSPSC Code:
12352102
PubChem Substance ID:

isotopic purity

98 atom % D

contains

~20% water as wetting agent

mp

106-108 °C (lit.)

mass shift

M+3

SMILES string

[2H]c1c([2H])c(c([2H])c(c1O)[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C6H4N2O5/c9-6-2-1-4(7(10)11)3-5(6)8(12)13/h1-3,9H/i1D,2D,3D

InChI key

UFBJCMHMOXMLKC-CBYSEHNBSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service