Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

42195

Sigma-Aldrich

3,4-Dinitrophenol

moistened with water, ≥97.0% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$163.00
5 G
$559.00

$163.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$163.00
5 G
$559.00

About This Item

Empirical Formula (Hill Notation):
C6H4N2O5
CAS Number:
Molecular Weight:
184.11
Beilstein:
1969398
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$163.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥97.0% (HPLC)

quality

moistened with water

contains

~20% water as stabilizer

mp

130-135 °C

solubility

dioxane: soluble 1 g/10 mL

functional group

nitro

SMILES string

Oc1ccc(c(c1)[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C6H4N2O5/c9-4-1-2-5(7(10)11)6(3-4)8(12)13/h1-3,9H

InChI key

AKLOLDQYWQAREW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,4-Dinitrophenol is a nitrophenol derivative.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Expl. 1.1 - STOT RE 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Angeles Camacho-Ruiz et al.
Analytical methods : advancing methods and applications, 12(32), 4048-4057 (2020-08-07)
A continuous spectrophotometric assay for the screening of PHB depolymerase activity in microtiter plates was developed. We evaluated crystalline PHB in the suspension and coated it with the addition of a pH indicator to detect the breakage of the ester

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service