Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

570966

Sigma-Aldrich

N-[1-(S)-(+)-Ethoxycarbonyl-3-phenylpropyl]-L-alanyl carboxyanhydride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19NO5
CAS Number:
Molecular Weight:
305.33
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

optical activity

[α]20/D +36.5°, c = 1 in chloroform

reaction suitability

reaction type: solution phase peptide synthesis

mp

69-73 °C (lit.)

application(s)

peptide synthesis

SMILES string

CCOC(=O)[C@H](CCc1ccccc1)N2[C@@H](C)C(=O)OC2=O

InChI

1S/C16H19NO5/c1-3-21-15(19)13(10-9-12-7-5-4-6-8-12)17-11(2)14(18)22-16(17)20/h4-8,11,13H,3,9-10H2,1-2H3/t11-,13-/m0/s1

InChI key

GFZFELCFSBCPDB-AAEUAGOBSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service