Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T1820002

Trapidil impurity A

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

7-Hydroxy-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine, 5-Methyl[1,2,4]triazolo[1,5-a]pyrimidin-7-ol, 6-Methyl-4-hydroxy-1,3,3a,7-tetraazaindene, 7-Hydroxy-5-methyl-1,3,4-triazaindolizine, NSC 32071, NSC 511493

Sign Into View Organizational & Contract Pricing

Select a Size

20 MG
$169.00

$169.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
20 MG
$169.00

About This Item

Empirical Formula (Hill Notation):
C6H6N4O
CAS Number:
Molecular Weight:
150.14
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$169.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

trapidil

manufacturer/tradename

EDQM

mp

280-283 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cc1cc(O)n2ncnc2n1

InChI

1S/C6H6N4O/c1-4-2-5(11)10-6(9-4)7-3-8-10/h2-3,11H,1H3

InChI key

ZUIVNYGZFPOXFW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Reactant for the synthesis of:
  • Ruthenium(II)-Hmtpo complexes
  • Dihydroorotate dehydrogenase inhibitors with antimalarial activity

Reactant for the Vilsmeier reaction of conjugated carbocycles and heterocycles

Investigations of the pharmacological activity caused by binding to HIV TAR RNA

Additive to study the space charge layer in silver bromide microcrystals

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service