Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

562572

Sigma-Aldrich

Cesium carbonate

Cabot high-purity grade

Synonym(s):

Carbonic acid dicesium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cs2CO3
CAS Number:
Molecular Weight:
325.82
Beilstein:
4546405
EC Number:
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:

grade

Cabot high-purity grade

mp

610 °C (dec.) (lit.)

SMILES string

[Cs+].[Cs+].[O-]C([O-])=O

InChI

1S/CH2O3.2Cs/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

FJDQFPXHSGXQBY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for:
  • Aerobic oxidation of primary alcohols and of arylalkanes to aryl ketones.†
  • 1,4-addition reactions

Reagent for:
  • Synthesis of phosphazene derivatives
  • Constrictive binding interactions
  • Intramolecular C-N cross coupling reactions
  • Dehydrohalogenative polycondensation
Promotes the efficient O-alkylation of alcohols to form mixed alkyl carbonates.

Legal Information

Product of Cabot Performance Materials

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 2 - STOT RE 2 Oral

Target Organs

Kidney,Adrenal gland,Testes

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, 3617-3617 (2006)
Aylin Uslu, et al
Inorgorganica Chimica Acta, 363, 3721-3726 (2010)
Xiao-Jin Wu, et al
Advanced Synthesis & Catalysis, 351, 3150-3156 (2009)
Yeon Sil Park, et al
Tetrahedron Letters, 51, 3956-3959 (2010)
Qifeng Wang et al.
Journal of the American Chemical Society, 132(33), 11420-11421 (2010-08-06)
Dehydrohalogenative polycondensation of 2-bromo-3-hexylthiophene was successful with Herrmann's catalyst and tris(2-dimethylaminophenyl)phosphine as catalyst precursors, giving head-to-tail poly(3-hexylthiophene) (HT-P3HT) with high molecular weight (M(n) = 30,600, M(w)/M(n) = 1.60) and high regioregularity (98%) in almost quantitative yield (99%).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service