MilliporeSigma
  • Palladium-catalyzed dehydrohalogenative polycondensation of 2-bromo-3-hexylthiophene: an efficient approach to head-to-tail poly(3-hexylthiophene).
Palladium-catalyzed dehydrohalogenative polycondensation of 2-bromo-3-hexylthiophene: an efficient approach to head-to-tail poly(3-hexylthiophene).
Journal of the American Chemical Society (2010-08-06)
Qifeng Wang, Ryo Takita, Yuuta Kikuzaki, Fumiyuki Ozawa
ABSTRACT
Dehydrohalogenative polycondensation of 2-bromo-3-hexylthiophene was successful with Herrmann's catalyst and tris(2-dimethylaminophenyl)phosphine as catalyst precursors, giving head-to-tail poly(3-hexylthiophene) (HT-P3HT) with high molecular weight (M(n) = 30,600, M(w)/M(n) = 1.60) and high regioregularity (98%) in almost quantitative yield (99%).
Sigma-Aldrich
Cesium carbonate, purum p.a., ≥98.0% (T)
Sigma-Aldrich
Cesium carbonate, 99.9% trace metals basis
Sigma-Aldrich
Cesium carbonate, ReagentPlus®, 99%
Sigma-Aldrich
trans-Bis(acetato)bis[o-(di-o-tolylphosphino)benzyl]dipalladium(II), 98%