Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

554286

Sigma-Aldrich

Mesitylacetic acid

97%

Synonym(s):

2,4,6-Trimethylphenylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3C6H2CH2CO2H
CAS Number:
Molecular Weight:
178.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

167-171 °C (lit.)

SMILES string

Cc1cc(C)c(CC(O)=O)c(C)c1

InChI

1S/C11H14O2/c1-7-4-8(2)10(6-11(12)13)9(3)5-7/h4-5H,6H2,1-3H3,(H,12,13)

InChI key

CQWMQAKKAHTCSC-UHFFFAOYSA-N

General description

α2-Chloroisodurene and mesitylacetonitrile are formed as intermediates during the synthesis of mesitylacetic acid. Mesitylacetic acid can be synthesized from mesitylene.

Application

Mesitylacetic acid may be used to synthesize 2,4,6-trimethvlbenzyl mesitylacetate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reaction Paths in the Kolbe Synthesis.
Rand L and Mohar AF.
The Journal of Organic Chemistry, 30(11), 3885-3888 (1965)
Synthesis of Mesitylacetic Acid.
Li H, et al.
Jing Xi Hua Gong Zhong Jian Ti / Fine Chemical Intermediates, 1, 009-009 (2010)
Mesitylacetic acid.
Fuson RC and Rabjohn N.
Organic Syntheses, 69-69 (1955)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service