Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

273023

Sigma-Aldrich

3,5-Di-tert-butylbenzoic acid

99%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$309.00

$309.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$309.00

About This Item

Linear Formula:
[(CH3)3C]2C6H3CO2H
CAS Number:
Molecular Weight:
234.33
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$309.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

solid

mp

173-175 °C (lit.)

SMILES string

CC(C)(C)c1cc(cc(c1)C(C)(C)C)C(O)=O

InChI

1S/C15H22O2/c1-14(2,3)11-7-10(13(16)17)8-12(9-11)15(4,5)6/h7-9H,1-6H3,(H,16,17)

InChI key

NCTSLPBQVXUAHR-UHFFFAOYSA-N

Application

3,5-Di-tert-butylbenzoic acid was used in the synthesis of 5,5-di-tert-butylanilinium sulphate[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

THE SYNTHESIS AND SUBSTITUTION REACTIONS OF 3, 5-DI-tert-BUTYLPHENOL.
Canadian Journal of Chemistry, 41(7), 1653-1656 (1963)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service