Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

548030

Sigma-Aldrich

3-Bromo-4-methylpyridine

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6BrN
CAS Number:
Molecular Weight:
172.02
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

refractive index

n20/D 1.56 (lit.)

bp

199-200 °C (lit.)

density

1.549 g/mL at 25 °C (lit.)

functional group

bromo

SMILES string

Cc1ccncc1Br

InChI

1S/C6H6BrN/c1-5-2-3-8-4-6(5)7/h2-4H,1H3

InChI key

GSQZOLXWFQQJHJ-UHFFFAOYSA-N

Application

3-Bromo-4-methylpyridine may be used as a building block in the preparation of:
  • substituted 4-(2,2-diphenylethyl)pyridine-N-oxides for use as potent phosphodiesterase type 4 (PDE4) inhibitors[1]
  • benzodiazepine site ligands bearing tricyclic pyridone moiety for human GABAA receptor[2]
  • a novel isomer of ascididemin[3]
  • 3-bromopyridine-4-carbonitrile[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

174.9 °F - closed cup

Flash Point(C)

79.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Richard Frenette et al.
Bioorganic & medicinal chemistry letters, 12(20), 3009-3013 (2002-09-25)
A detailed SAR study directed toward the optimization of pharmacokinetic parameters for analogues of L-791,943 is reported. The introduction of a soft metabolic site on this structure permitted the identification of L-826,141 as a potent phosphodiesterase type 4 (PDE4) inhibitor
James Crawforth et al.
Bioorganic & medicinal chemistry letters, 14(7), 1679-1682 (2004-03-18)
A series of tricyclic pyridones has been evaluated as benzodiazepine site ligands with functional selectivity for the alpha(3) over the alpha(1) containing subtype of the human GABA(A) receptor ion channel. This investigation led to the identification of a high affinity
Condensed heteroaromatic ring systems. XV. Synthesis of pyranopyridinones from halopyridinecarbonitriles.
Sakamoto T, et al.
Chemical & Pharmaceutical Bulletin, 36(5), 1890-1894 (1988)
Ida Nymann Petersen et al.
Chemical communications (Cambridge, England), 48(72), 9092-9094 (2012-08-07)
A new and convergent synthesis of ascididemin is presented. Using an anionic cascade ring closure as the key step, this natural product is obtained in 45% overall yield in just 6 steps starting from 2'-fluoroacetophenone. This new approach was extended

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service